Would you like to log in again?
Exocell Nephrat II, Rat Albumin ELISA Assay, Ethos Biosciences
Supplier: Ethos Biosciences
NR002
MSPP-NR002EA 1272.31 USD InStock
MSPP-NR002
Exocell Nephrat II, Rat Albumin ELISA Assay, Ethos Biosciences
Assays ELISAs
Direct competitive antibody capture
  • Specimen required: Urine, 10 µL
Precision: Intrassay and interassay precision for samples within useful range of the assay have a C.V. ≦ 10% of the mean
2 x 96 well plates. 80 sample wells per plate. 16 standard wells per plate. 160 total sample wells. 160 total samples or 80 in duplicate (recommended).
Ordering information: Our kits provide all of the necessary components, including overages, to conduct your assays with confidence.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty